Anti-FUT1 Goat Polyclonal Antibody

Supplier: Biorbyt
BIRBORB22497-100EA 372 CHF
BIRBORB22497-100
Anti-FUT1 Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: FUT1 (fucosyltransferase 1 (galactoside 2-alpha-L-fucosyltransferase, H blood group))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR