Anti-IgM Rabbit Polyclonal Antibody

Lieferant: Biorbyt
BIRBORB22453-1EA 459 CHF
BIRBORB22453-1
Anti-IgM Rabbit Polyclonal Antibody
Antikörper
Type: Secondary
Antigen: IgM (Goat) (goat IgM)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR