Anti-IgG2a Sheep Polyclonal Antibody

Supplier: Biorbyt
BIRBORB22445-1EA 459 CHF
BIRBORB22445-1
Anti-IgG2a Sheep Polyclonal Antibody
Antibodies
Type: Secondary
Antigen: IgG2a (Mouse) (mouse IgG2a)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR