Anti-IgG1 Sheep Polyclonal Antibody

Lieferant: Biorbyt
BIRBORB22424-1EA 459 CHF
BIRBORB22424-1
Anti-IgG1 Sheep Polyclonal Antibody
Antikörper
Type: Secondary
Antigen: IgG1 (Bovine) (bovine IgG1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR