Anti-IgA Rabbit Polyclonal Antibody

Supplier: Biorbyt
BIRBORB22354-1EA 419 CHF
BIRBORB22354-1
Anti-IgA Rabbit Polyclonal Antibody
Antibodies
Type: Secondary
Antigen: IgA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR