Anti-IgE Mouse Monoclonal Antibody (Biotin)

Supplier: Biorbyt
BIRBORB22243-0.2EA 801 CHF
BIRBORB22243-0.2
Anti-IgE Mouse Monoclonal Antibody (Biotin)
Antibodies
Type: Secondary
Antigen: IgE
Clonality: Monoclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR