Anti-IgG2 Mouse Monoclonal Antibody

Supplier: Biorbyt
BIRBORB22211-0.5EA 555 CHF
BIRBORB22211-0.5
Anti-IgG2 Mouse Monoclonal Antibody
Antibodies
Type: Secondary
Antigen: IgG2
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR