Anti-C3C Goat Polyclonal Antibody

Lieferant: Biorbyt
BIRBORB22119-10EA 0 CHF
BIRBORB22119-10
Anti-C3C Goat Polyclonal Antibody
Antikörper
Type: Secondary
Antigen: Monkey C3C
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR