Anti-C3C Goat Polyclonal Antibody (Biotin)

Supplier: Biorbyt
BIRBORB22114-1EA 518 CHF
BIRBORB22114-1
Anti-C3C Goat Polyclonal Antibody (Biotin)
Antibodies
Type: Secondary
Antigen: Monkey C3C
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR