Anti- IgG2a Polyclonal Antibody (Unconjugated)

Supplier: Biorbyt
BIRBORB22109-10EA 666 CHF
BIRBORB22109-10
Anti- IgG2a Polyclonal Antibody (Unconjugated)
Antibodies
Type: Secondary
Antigen: IgG2a (Mouse) (mouse IgG2a)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: None
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR