Anti-IgG2a Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Biorbyt
BIRBORB22104-1EA 666 CHF
BIRBORB22104-1
Anti-IgG2a Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Type: Secondary
Antigen: IgG2a (Mouse) (mouse IgG2a)
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: None
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR