Anti-Secretory ComponentSecretory Component Rabbit Polyclonal Antibody

Supplier: Biorbyt
BIRBORB22019-1EA 555 CHF
BIRBORB22019-1
Anti-Secretory ComponentSecretory Component Rabbit Polyclonal Antibody
Antibodies
Type: Secondary
Antigen: Secretory Component
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR