Anti-C1Q Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: Biorbyt
BIRBORB21949-1EA 518 CHF
BIRBORB21949-1
Anti-C1Q Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Type: Secondary
Antigen: C1Q
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Goat
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR