Anti-Transferrin Goat Polyclonal Antibody

Supplier: Biorbyt
BIRBORB21919-1EA 459 CHF
BIRBORB21919-1
Anti-Transferrin Goat Polyclonal Antibody
Antibodies
Type: Secondary
Antigen: Transferrin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR