Anti-C3C Goat Polyclonal Antibody

Supplier: Biorbyt
BIRBORB21904-1EA 459 CHF
BIRBORB21904-1
Anti-C3C Goat Polyclonal Antibody
Antibodies
Type: Secondary
Antigen: Dog C3C
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR