Anti-IgG Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))

Supplier: Biorbyt
BIRBORB21829-1EA 518 CHF
BIRBORB21829-1
Anti-IgG Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: TRITC (Tetramethylrhodamine Isothiocyanante)
Epitope:
Host:
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR