Anti-Lactoferrin Goat Polyclonal Antibody (Biotin)

Supplier: Biorbyt
BIRBORB21683-1EA 666 CHF
BIRBORB21683-1
Anti-Lactoferrin Goat Polyclonal Antibody (Biotin)
Antibodies
Type: Secondary
Antigen: Monkey Lactoferrin
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR