Anti-IgG1/IgG2a/IgG2b/IgG3 Rabbit Polyclonal Antibody (Biotin)

Lieferant: Biorbyt
BIRBORB21671-1EA 666 CHF
BIRBORB21671-1
Anti-IgG1/IgG2a/IgG2b/IgG3 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Secondary
Antigen: Mouse IgG1 IgG2A IgG2B IgG3
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR