Anti-IgD Goat Polyclonal Antibody (Biotin)

Supplier: Biorbyt
BIRBORB21640-1EA 0 CHF
BIRBORB21640-1
Anti-IgD Goat Polyclonal Antibody (Biotin)
Antibodies
Type: Secondary
Antigen: Mouse IgD
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR