Anti-IgM Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: Biorbyt
BIRBORB21609-1EA 518 CHF
BIRBORB21609-1
Anti-IgM Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
Type: Secondary
Antigen: IgM (Rat) (rat IgM)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Goat
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR