Anti-IgG1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: Biorbyt
BIRBORB21599-1EA 666 CHF
BIRBORB21599-1
Anti-IgG1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Type: Secondary
Antigen: IgG1
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR