Anti-IgA Pig Polyclonal Antibody

Lieferant: Biorbyt
BIRBORB21580-10EA 422 CHF
BIRBORB21580-10
Anti-IgA Pig Polyclonal Antibody
Antikörper
Type: Secondary
Antigen: IgA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Pig
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR