Anti-IgG Goat Polyclonal Antibody

Supplier: Biorbyt
BIRBORB21436-10EA 422 CHF
BIRBORB21436-10
Anti-IgG Goat Polyclonal Antibody
Antibodies
Type: Secondary
Antigen: Monkey IgG
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR