Anti-IgM Goat Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))

Supplier: Biorbyt
BIRBORB21433-1EA 666 CHF
BIRBORB21433-1
Anti-IgM Goat Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))
Antibodies
Type: Secondary
Antigen: IgM
Clonality: Polyclonal
Clone:
Conjugation: TRITC (Tetramethylrhodamine Isothiocyanante)
Epitope:
Host: Goat
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR