Anti-FC Mouse Monoclonal Antibody

Supplier: Biorbyt
BIRBORB21286-100EA 424 CHF
BIRBORB21286-100
Anti-FC Mouse Monoclonal Antibody
Antibodies
Western Blot: 1:1000

Type: Primary
Antigen: FC
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR