Anti-TRITC Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Biorbyt
BIRBORB21197-0.2EA 0 CHF
BIRBORB21197-0.2
Anti-TRITC Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Type: Primary
Antigen: TRITC
Clonality: monoclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR