Anti-Enolase Sheep Polyclonal Antibody

Supplier: Biorbyt
BIRBORB21135-10EA 422 CHF
BIRBORB21135-10
Anti-Enolase Sheep Polyclonal Antibody
Antibodies
Type: Primary
Antigen: Enolase
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: none
Reactivity: Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR