Anti-Formate Dehydrogenase Rabbit Polyclonal Antibody

Supplier: Biorbyt
BIRBORB21126-10EA 422 CHF
BIRBORB21126-10
Anti-Formate Dehydrogenase Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: Formate Dehydrogenase
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Yeast
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR