Anti-Dextranase Rabbit Polyclonal Antibody (Biotin)

Supplier: Biorbyt
BIRBORB21049-1EA 518 CHF
BIRBORB21049-1
Anti-Dextranase Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type: Primary
Antigen: Dextranase
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Bacteria
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR