Anti-Oxalate Oxidase Rabbit Polyclonal Antibody

Supplier: Biorbyt
BIRBORB20899-10EA 422 CHF
BIRBORB20899-10
Anti-Oxalate Oxidase Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: Oxalate Oxidase
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR