Anti-Ribonuclease T1 Rabbit Polyclonal Antibody (Biotin)

Supplier: Biorbyt
BIRBORB20852-1EA 518 CHF
BIRBORB20852-1
Anti-Ribonuclease T1 Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type: Primary
Antigen: Ribonuclease T1
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR