Anti-Amyloglucosidase Rabbit Polyclonal Antibody

Supplier: Biorbyt
BIRBORB20846-10EA 422 CHF
BIRBORB20846-10
Anti-Amyloglucosidase Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: Amyloglucosidase
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR