Anti-GAPDH None Polyclonal Antibody

Supplier: Biorbyt
BIRBORB20816-1EA 666 CHF
BIRBORB20816-1
Anti-GAPDH None Polyclonal Antibody
Antibodies
Type: Primary
Antigen: GAPDH (glyceraldehyde-3-phosphate dehydrogenase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: None
Isotype: none
Reactivity: Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR