Anti-Uridine-5'-Diphosphoglucose Dehydrogenase Rabbit Polyclonal Antibody

Supplier: Biorbyt
BIRBORB20721-1EA 666 CHF
BIRBORB20721-1
Anti-Uridine-5'-Diphosphoglucose Dehydrogenase Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: Uridine-5'-Diphosphoglucose Dehydrogenase
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR