Anti-Glycerokinase Rabbit Polyclonal Antibody

Supplier: Biorbyt
BIRBORB20691-1EA 666 CHF
BIRBORB20691-1
Anti-Glycerokinase Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: Glycerokinase
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Bacillus ssp.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR