Anti-AFP None Polyclonal Antibody (FITC (Fluorescein))

Supplier: Biorbyt
BIRBORB20676-1EA 666 CHF
BIRBORB20676-1
Anti-AFP None Polyclonal Antibody (FITC (Fluorescein))
Antibodies
Type: Primary
Antigen: AFP (alpha-fetoprotein)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: None
Isotype: none
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR