Anti-IgM Rabbit Polyclonal Antibody

Supplier: Biorbyt
BIRBORB20668-10EA 586 CHF
BIRBORB20668-10
Anti-IgM Rabbit Polyclonal Antibody
Antibodies
Type: Secondary
Antigen: IgM (Bovine) (bovine IgM)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR