Anti-DNAH8 Goat Polyclonal Antibody

Supplier: Biorbyt
BIRBORB20645-100EA 372 CHF
BIRBORB20645-100
Anti-DNAH8 Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: DNAH8 (dynein, axonemal, heavy chain 8)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR