Anti-NTN4 Goat Polyclonal Antibody

Lieferant: Biorbyt
BIRBORB20616-100EA 559 CHF
BIRBORB20616-100
Anti-NTN4 Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: NTN4 (netrin 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR