Anti-IMP3 Goat Polyclonal Antibody

Supplier: Biorbyt
BIRBORB20607-100EA 559 CHF
BIRBORB20607-100
Anti-IMP3 Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: IMP3 (IMP3, U3 small nucleolar ribonucleoprotein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR