Anti-ALDH1B1 Goat Polyclonal Antibody

Supplier: Biorbyt
BIRBORB20589-100EA 559 CHF
BIRBORB20589-100
Anti-ALDH1B1 Goat Polyclonal Antibody
Antibodies
ELISA: 1:4000; Western Blot: 0.3-1 ?g/ml

Type: Primary
Antigen: ALDH1B1 (aldehyde dehydrogenase 1 family, member B1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR