Anti-PEX26 Goat Polyclonal Antibody

Supplier: Biorbyt
BIRBORB20565-100EA 559 CHF
BIRBORB20565-100
Anti-PEX26 Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PEX26 (peroxisomal biogenesis factor 26)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR