Anti-POU3F3 Goat Polyclonal Antibody

Supplier: Biorbyt
BIRBORB20546-100EA 559 CHF
BIRBORB20546-100
Anti-POU3F3 Goat Polyclonal Antibody
Antibodies
ELISA: 1:4000; Western Blot: 1-3 ?g/ml

Type: Primary
Antigen: POU3F3 (POU class 3 homeobox 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR