Anti-EXT2 Goat Polyclonal Antibody

Lieferant: Biorbyt
BIRBORB20499-100EA 0 CHF
BIRBORB20499-100
Anti-EXT2 Goat Polyclonal Antibody
Antikörper
ELISA: 1:8000; Western Blot: 1 ?g/ml

Type: Primary
Antigen: EXT2 (exostosin glycosyltransferase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR