Anti-FREM2 Goat Polyclonal Antibody

Supplier: Biorbyt
BIRBORB20490-100EA 372 CHF
BIRBORB20490-100
Anti-FREM2 Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: FREM2 (FRAS1 related extracellular matrix protein 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR