Anti-TOMM40 Goat Polyclonal Antibody

Supplier: Biorbyt
BIRBORB20470-100EA 559 CHF
BIRBORB20470-100
Anti-TOMM40 Goat Polyclonal Antibody
Antibodies
ELISA: 1:16000; Western Blot: 1-3 ?g/ml

Type: Primary
Antigen: TOMM40 (translocase of outer mitochondrial membrane 40 homolog (yeast))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR