Anti-CRISP2 Goat Polyclonal Antibody

Supplier: Biorbyt
BIRBORB20468-100EA 559 CHF
BIRBORB20468-100
Anti-CRISP2 Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CRISP2 (cysteine-rich secretory protein 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR