Anti-UBP1 Goat Polyclonal Antibody

Lieferant: Biorbyt
BIRBORB20441-100EA 559 CHF
BIRBORB20441-100
Anti-UBP1 Goat Polyclonal Antibody
Antikörper
ELISA: 1:2000; Western Blot: 2 ?g/ml

Type: Primary
Antigen: UBP1 (upstream binding protein 1 (LBP-1a))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR