Anti-PCSK5 Goat Polyclonal Antibody

Supplier: Biorbyt
BIRBORB20386-100EA 372 CHF
BIRBORB20386-100
Anti-PCSK5 Goat Polyclonal Antibody
Antibodies
ELISA: 1:8000; Western Blot: 1 ?g/ml

Type: Primary
Antigen: PCSK5 (proprotein convertase subtilisin/kexin type 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR