Anti-APOL6 Goat Polyclonal Antibody

Supplier: Biorbyt
BIRBORB20207-100EA 559 CHF
BIRBORB20207-100
Anti-APOL6 Goat Polyclonal Antibody
Antibodies
ELISA: 1:2000; Western Blot: 1-3 ?g/ml

Type: Primary
Antigen: APOL6 (apolipoprotein L, 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR